CAS RN 1347815-42-2
CAS RN 1347815-42-2
IUPAC Name:
1-(cyanomethyl)-2-(3-cyanophenyl)-1H-imidazole-4,5-dicarbonitrile
Synonyms:
1-(cyanomethyl)-2-(3-cyanophenyl)imidazole-4,5-dicarbonitrile
Molecular Weight (g/mol):
258.24
Molecular Formula:
C14H6N6
InChi Key:
KQHKTLGFDJPVEP-UHFFFAOYSA-N
SMILES:
N#CCN1C(=NC(C#N)=C1C#N)C1=CC=CC(=C1)C#N
1
–
1
of
1
results
1
1-Cyanomethyl-2-(3-cyanophenyl)-4,5-dicyanoimidazole, 97%, Thermo Scientific™
CAS: 1347815-42-2 Molecular Formula: C14H6N6 Molecular Weight (g/mol): 258.244 MDL Number: MFCD20265160 InChI Key: KQHKTLGFDJPVEP-UHFFFAOYSA-N Synonym: 1-cyanomethyl-2-3-cyanophenyl-4,5-dicyanoimidazole,1-cyanomethyl-2-3-cyanophenyl imidazole-4,5-dicarbonitrile PubChem CID: 73995593 IUPAC Name: 1-(cyanomethyl)-2-(3-cyanophenyl)imidazole-4,5-dicarbonitrile SMILES: C1=CC(=CC(=C1)C2=NC(=C(N2CC#N)C#N)C#N)C#N