accessibility menu, dialog, popup

UserName

CAS RN 134444-47-6

NNOHOHOFNNNH2NH2

CAS RN 134444-47-6

IUPAC Name: 5-(2,6-diamino-9H-purin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
Synonyms: 5-(2,6-diaminopurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
Molecular Weight (g/mol): 284.25
Molecular Formula: C10H13FN6O3
InChi Key: MHWHYOFBOWTAHZ-UHFFFAOYNA-N
SMILES: NC1=NC(N)=C2N=CN(C3OC(CO)C(O)C3F)C2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results