accessibility menu, dialog, popup

UserName

CAS RN 134-63-4

ClCH3N(S)(R)O(S)HOH

CAS RN 134-63-4

IUPAC Name: hydrogen 2-[(2R,6S)-6-[(2S)-2-hydroxy-2-phenylethyl]-1-methylpiperidin-2-yl]-1-phenylethan-1-one chloride
Synonyms: hydrogen (-)-lobeline chloride
Molecular Weight (g/mol): 373.92
Molecular Formula: C22H28ClNO2
InChi Key: MKMYPTLXLWOUSO-NFQNBQCWSA-N
SMILES: [H+].[Cl-].CN1[C@H](C[C@H](O)C2=CC=CC=C2)CCC[C@@H]1CC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results