missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lobeline hydrochloride, MedChemExpress
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Lobeline hydrochloride, a nicotinic receptor agonist, acting as a potent antagonist at both α3β2 and α4β2 neuronal nicotinic receptor subtypes.
Specifications
Specifications
| Chemical Name or Material | Lobeline hydrochloride |
| Physical Form | Solid |
| CAS | 134-63-4 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H28ClNO2 |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Synonym | α-Lobeline hydrochlorideL-Lobeline hydrochloride |
| Solubility Information | H2O : 6 mg/mL (16.05 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction