CAS RN 13389-03-2
IUPAC Name:
2-amino-8-bromo-9-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-3H-purin-6-one
Synonyms:
2-amino-8-bromo-9-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-purin-6-one
Molecular Weight (g/mol):
346.14
Molecular Formula:
C10H12BrN5O4
InChi Key:
MKDXZFVCXWXGBQ-UHFFFAOYNA-N
SMILES:
NC1=NC(=O)C2=C(N1)N(C1CC(O)C(CO)O1)C(Br)=N2
1
–
3
of
3
results
2
8-Bromo-2’-deoxyguanosine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
8-Bromo-2’-deoxyguanosine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.