missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-2’-deoxyguanosine, TRC

Product Code. 30343082
Change view
Click to view available options
Quantity:
25 mg
1 g
Unit Size:
1g
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30343082 1 g 1g
30334637 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30343082 Supplier Toronto Research Chemicals Supplier No. TRCB6828351G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 8-Bromo-2'-deoxyguanosine
CAS 13389-03-2
Molecular Formula C10 H12 Br N5 O4
Quantity 1 g
Synonym Guanosine, 8-bromo-2'-deoxy-, 8-Bromo-2'-deoxyguanosine, NSC 105830
InChI Formula InChI=1S/C10H12BrN5O4/c11-9-13-6-7(14-10(12)15-8(6)19)16(9)5-1-3(18)4(2-17)20-5/h3-5,17-18H,1-2H2,(H3,12,14,15,19)/t3-,4+,5+/m0/s1
SMILES NC1=Nc2c(nc(Br)n2[C@H]3C[C@H](O)[C@@H](CO)O3)C(=O)N1
IUPAC Name 2-amino-8-bromo-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1H-purin-6-one
Molecular Weight (g/mol) 346.14
Formula Weight 345.0073
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.