CAS RN 1332331-08-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ GSK 2194069
Potent human fatty acid synthase (hFASN) inhibitor
2
GSK2194069, MedChemExpress
MedChemExpress GSK2194069 is a potent inhibitor of β-ketoyl reductase (KR) of fatty acid synthase (FAS), with an IC50 value of 7.7 nM. GSK2194069 shows specifically inhibitory effect on FAS expressing cancer cells, by acting potent efficacy on acetoacetyl-CoA, NADPH with IC50 or Ki values of 4.8 nM and 5.6 nM, respectively.