missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK2194069, MedChemExpress
Description
GSK2194069 is a potent inhibitor of β-ketoyl reductase (KR) of fatty acid synthase (FAS), with an IC50 value of 7.7 nM. GSK2194069 shows specifically inhibitory effect on FAS expressing cancer cells, by acting potent efficacy on acetoacetyl-CoA, NADPH with IC50 or Ki values of 4.8 nM and 5.6 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | GSK2194069 |
| Physical Form | Solid |
| CAS | 1332331-08-4 |
| Color | Earth Yellow |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C25H24N4O3 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : ≥ 100 mg/mL (233.38 mM) |
| SMILES | O=C(N1C[C@H](CC2=NNC(N2C3=CC=C(C4=CC=C(OC=C5)C5=C4)C=C3)=O)CC1)C6CC6 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?