accessibility menu, dialog, popup

UserName

CAS RN 132980-99-5

H2NOFF

CAS RN 132980-99-5

IUPAC Name: 3,5-difluorobenzamide
Synonyms: 3,5-difluorobenzamide
Molecular Weight (g/mol): 157.12
Molecular Formula: C7H5F2NO
InChi Key: CGOIBYYWOVRGGV-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC(F)=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results