accessibility menu, dialog, popup

UserName

CAS RN 1323966-24-0

CH3OFOClCl

CAS RN 1323966-24-0

IUPAC Name: 4-chloro-2-fluoro-3-methoxybenzoyl chloride
Synonyms: 4-chloro-2-fluoro-3-methoxybenzoyl chloride
Molecular Weight (g/mol): 223.02
Molecular Formula: C8H5Cl2FO2
InChi Key: FPFZZDWTMMQEHB-UHFFFAOYSA-N
SMILES: COC1=C(Cl)C=CC(C(Cl)=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results