accessibility menu, dialog, popup

UserName

CAS RN 130964-39-5

NSOOHNNHBrH2OHClHCl(E/Z)

CAS RN 130964-39-5

IUPAC Name: N-(2-{[3-(4-bromophenyl)prop-2-en-1-yl]amino}ethyl)isoquinoline-5-sulfonamide hydrate dihydrochloride
Synonyms: N-(2-{[3-(4-bromophenyl)prop-2-en-1-yl]amino}ethyl)isoquinoline-5-sulfonamide hydrate dihydrochloride
Molecular Weight (g/mol): 537.29
Molecular Formula: C20H24BrCl2N3O3S
InChi Key: GKFFJFGBWAGAFD-UHFFFAOYSA-N
SMILES: O.Cl.Cl.BrC1=CC=C(C=CCNCCNS(=O)(=O)C2=C3C=CN=CC3=CC=C2)C=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results