missing translation for 'onlineSavingsMsg'
Learn More

H-89 dihydrochloride, MedChemExpress

Product Code. 16466948
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466948 10 mg 10mg
16457026 100 mg 100mg
16443007 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16466948 Supplier Medchem Express Supplier No. HY15979A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress H-89 dihydrochloride is a potent and selective inhibitor of protein kinase A (PKA) with an IC50 of 48 nM and has weak inhibition on PKG, PKC, Casein Kinase.

H-89 dihydrochloride is a potent and selective inhibitor of protein kinase A (PKA) with an IC50 of 48 nM and has weak inhibition on PKG, PKC, Casein Kinase.

Specifications

Chemical Name or Material H-89 dihydrochloride
CAS 130964-39-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H22BrCl2N3O2S
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (192.57 mM; Need ultrasonic) ∣H2O : 5 mg/mL (9.63 mM; ultrasonic and warming and heat to 80°C)
SMILES O=S(C1=CC=CC2=C1C=CN=C2)(NCCNC/C=C/C3=CC=C(Br)C=C3)=O.Cl.Cl
Molecular Weight (g/mol) 519.28
Formula Weight 519.28
Percent Purity 98.71%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.