missing translation for 'onlineSavingsMsg'
Learn More

N-[2-(p-Bromocinnamylamino)ethyl]-5-isoquinoline Sulfonamide Dihydrochloride, TRC

Product Code. 30342528
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30342528 50 mg 50mg
30333201 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30342528 Supplier Toronto Research Chemicals Supplier No. TRCB68244550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[2-(p-Bromocinnamylamino)ethyl]-5-isoquinoline Sulfonamide Dihydrochloride
CAS 130964-39-5
Molecular Formula C20H22BrCl2N3O2S
Quantity 50 mg
Synonym N-[2-[[3-(4-Bromophenyl)-2-propen-1-yl]amino]ethyl]-5-isoquinolinesulfonamide Hydrochloride, H-89 Dihydrochloride
InChI Formula InChI=1S/C20H20BrN3O2S.2ClH/c21-18-8-6-16(7-9-18)3-2-11-22-13-14-24-27(25,26)20-5-1-4-17-15-23-12-10-19(17)20;;/h1-10,12,15,22,24H,11,13-14H2;2*1H/b3-2+;;
SMILES Cl.Cl.Brc1ccc(\C=C\CNCCNS(=O)(=O)c2cccc3cnccc23)cc1
IUPAC Name N-[2-[[(E)-3-(4-bromophenyl)prop-2-enyl]amino]ethyl]isoquinoline-5-sulfonamide;dihydrochloride
Molecular Weight (g/mol) 519.28
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.