missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Formyl-L-leucine [S-(E)]-1-(2-Nonenyl)dodecyl Ester, Mikromol™
Description
N-Formyl-L-leucine [S-(E)]-1-(2-Nonenyl)dodecyl Ester
Specifications
Specifications
| Chemical Name or Material | N-Formyl-L-leucine [S-(E)]-1-(2-Nonenyl)dodecyl Ester |
| CAS | 130676-63-0 |
| Analyte or Component Names | N-Formyl-L-leucine [S-(E)]-1-(2-Nonenyl)dodecyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C28 H53 N O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | N-Formyl-L-leucine [S-(E)]-1-(2-Nonenyl)dodecyl Ester (9Cl) |
| InChI Formula | InChI=1S/C28H53NO3/c1-5-7-9-11-13-14-16-18-20-22-26(21-19-17-15-12-10-8-6-2)32-28(31)27(29-24-30)23-25(3)4/h17,19,24-27H,5-16,18,20-23H2,1-4H3,(H,29,30)/b19-17+/t26-,27+/m1/s1 |
| SMILES | CCCCCCCCCCC[C@@H](C\C=C\CCCCCC)OC(=O)[C@H](CC(C)C)NC=O |
| IUPAC Name | [(E,10S)-henicos-7-en-10-yl] (2S)-2-formamido-4-methylpentanoate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?