CAS RN 1305116-67-9
CAS RN 1305116-67-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tocris Bioscience™ PF 04671536 hydrochloride
Potent and selective PDE8B/8A inhibitor