accessibility menu, dialog, popup

UserName

CAS RN 1291-47-0

FeCH3CH3

CAS RN 1291-47-0

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 210.06
Molecular Formula: C12H10Fe
InChi Key: SNGSYGKLJUGRID-UHFFFAOYSA-N
SMILES: C[c-]1cccc1.C[C-]12[C-]3[C-]4[C-]5[C-]1[Fe]2345
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results