accessibility menu, dialog, popup

UserName

CAS RN 1276121-88-0

CH3HNONHSNNH3C

CAS RN 1276121-88-0

IUPAC Name: 15-methyl-5-(6-methylpyridin-3-yl)-11-thia-6,14,17-triazatetracyclo[8.8.0.0²,⁷.0¹²,¹⁸]octadeca-1(10),2,4,6,8,12(18)-hexaen-13-one
Synonyms: 15-methyl-5-(6-methylpyridin-3-yl)-11-thia-6,14,17-triazatetracyclo[8.8.0.0²,⁷.0¹²,¹⁸]octadeca-1(10),2,4,6,8,12(18)-hexaen-13-one
Molecular Weight (g/mol): 374.46
Molecular Formula: C21H18N4OS
InChi Key: CMWRPDHVGMHLSZ-UHFFFAOYNA-N
SMILES: CC1CNC2=C(SC3=C2C2=CC=C(N=C2C=C3)C2=CN=C(C)C=C2)C(=O)N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results