missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 3644022
GREENER_CHOICE

Product Code. 18085092
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Product Code. Quantity unitSize
18085092 10 mg 10mg
1 options
This item is not returnable. View return policy

Product Code. 18085092

Brand: Tocris Bioscience™ 4279/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent MK2 inhibitor

Potent, ATP-competitive inhibitor of mitogen-activated protein kinase (MAPK)-activated protein kinase-2 (MK2) (IC50 = 5.2 nM; Ki = 3 nM). Inhibits tumor necrosis factor α (TNFα) production in U937 monocytic cells and peripheral blood mononuclear cells (PBMCs) (IC50 = 160 nM); exhibits oral efficacy in acute and chronic inflammatory models.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1276121-88-0
Quantity 10 mg
Molecular Formula C21H18N4OS
Formula Weight 374.46
Purity >98%
Molecular Weight (g/mol) 374.462
Chemical Name or Material PF 3644022
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.