accessibility menu, dialog, popup

UserName

CAS RN 1266124-50-8

SFFFFFOHONH2

CAS RN 1266124-50-8

IUPAC Name: 2-amino-3-[3-(pentafluoro-λ⁶-sulfanyl)phenyl]propanoic acid
Synonyms: 2-amino-3-[3-(pentafluoro-λ⁶-sulfanyl)phenyl]propanoic acid
Molecular Weight (g/mol): 291.24
Molecular Formula: C9H10F5NO2S
InChi Key: VZQSFTLJODGXCO-UHFFFAOYNA-N
SMILES: NC(CC1=CC(=CC=C1)S(F)(F)(F)(F)F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)