CAS RN 126544-47-6
CAS RN 126544-47-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Ciclesonide 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Ciclesonide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
3
Ciclesonide, MedChemExpress
MedChemExpress Ciclesonide (RPR251526) is a glucocorticoid with an potent anti-inflammatory activity. Ciclesonide can be used for asthma research.
4
Ciclesonide
Glucocorticoid antiasthmatic prodrug