missing translation for 'onlineSavingsMsg'
Learn More

Ciclesonide

Product Code. 18757016
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18757016 25 mg 25mg
18044112 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18757016 Supplier Tocris Bioscience Supplier No. 6357/25

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Glucocorticoid antiasthmatic prodrug

Ciclesonide is a glucocorticoid antiasthmatic prodrug; de-esterified in the lung to active metabolite. Anti-inflammatory. Also blocks replication of SARS-CoV-2 in Vero cells in vitro by targeting viral NSP-15 (EC90 = 6.3μM)..
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (11β,16α)-16,17-[[(R)-Cyclohexylmethylene]bis(oxy)]-11-hydroxy-21-(2-methyl-1-oxopropoxy)pregna-1,4-diene-3,20-dione
CAS 126544-47-6
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C32H44O7
Formula Weight Observed MW: 540.7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.