CAS RN 1261491-89-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Olumacostat glasaretil, MedChemExpress
MedChemExpress Olumacostat glasaretil is a small molecule inhibitor of acetyl coenzyme A carboxylase (ACC).