missing translation for 'onlineSavingsMsg'
Learn More

Olumacostat glasaretil, MedChemExpress

Product Code. 16477438
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
1mg
200mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477438 10 mg 10mg
16459917 5 mg 5mg
16451276 200 mg 200mg
16441719 1 mg 1mg
16423467 50 mg 50mg
16417416 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16477438 Supplier Medchem Express Supplier No. HY17641

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Olumacostat glasaretil is a small molecule inhibitor of acetyl coenzyme A carboxylase (ACC).

Olumacostat glasaretil is a small molecule inhibitor of acetyl coenzyme A carboxylase (ACC).

Specifications

Chemical Name or Material Olumacostat glasaretil
Physical Form Solid
CAS 1261491-89-7
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C26H43NO7
Solubility Information DMSO : 125 mg/mL (259.54 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(OCCCCCCCCCCCCCC)O1)OCC(N(CC(OCC)=O)C)=O
Molecular Weight (g/mol) 481.62
Formula Weight 481.62
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.