missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tolterodine tartrate, MedChemExpress
Description
Tolterodine Tartrate (Kabi-2234
Specifications
Specifications
| Chemical Name or Material | Tolterodine tartrate |
| CAS | 124937-52-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C26H37NO7 |
| Quantity | 50 mg |
| Synonym | Kabi-2234 PNU-200583E |
| Solubility Information | DMSO : 100 mg/mL (210.27 mM; Need ultrasonic) ∣H2O : 16.67 mg/mL (35.05 mM; Need ultrasonic) |
| SMILES | OC1=CC=C(C=C1[C@H](CCN(C(C)C)C(C)C)C2=CC=CC=C2)C.O=C([C@@H]([C@H](C(O)=O)O)O)O |
| Molecular Weight (g/mol) | 475.57 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?