accessibility menu, dialog, popup

UserName

CAS RN 123439-82-7

OOHOOOHOPtNNNN

CAS RN 123439-82-7

IUPAC Name: bis(carbonic acid) tetraamine platinum
Synonyms: bis(carbonic acid) tetraamine platinum
Molecular Weight (g/mol): 387.26
Molecular Formula: C2H16N4O6Pt
InChi Key: AGUMVCVDCJQCIQ-UHFFFAOYSA-N
SMILES: N.N.N.N.[Pt].OC(O)=O.OC(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results