CAS RN 1224887-10-8
CAS RN 1224887-10-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Tocris Bioscience™ GSK 2256098
Potent focal adhesion kinase (FAK) inhibitor
2
GSK2256098, MedChemExpress
MedChemExpress GSK2256098 is a selective FAK kinase inhibitor, which inhibits growth and survival of pancreatic ductal adenocarcinoma cells.