missing translation for 'onlineSavingsMsg'
Learn More

GSK2256098, MedChemExpress

Product Code. 16484346
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
2mg
50mg
5mg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16484346 100 mg 100mg
16453048 10 mg 10mg
16425787 5 mg 5mg
16419616 50 mg 50mg
16402596 25 mg 25mg
16400916 200 mg 200mg
16400026 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 16484346 Supplier Medchem Express Supplier No. HY100498

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GSK2256098 is a selective FAK kinase inhibitor, which inhibits growth and survival of pancreatic ductal adenocarcinoma cells.

GSK2256098 is a selective FAK kinase inhibitor, which inhibits growth and survival of pancreatic ductal adenocarcinoma cells.

Specifications

Chemical Name or Material GSK2256098
CAS 1224887-10-8
Color White
Purity Grade Notes Research
Molecular Formula C20H23ClN6O2
Quantity 100 mg
Solubility Information DMSO : ≥ 30 mg/mL (72.31 mM)
SMILES O=C(NOC)C1=CC=CC=C1NC2=CC(NC3=CC(C)=NN3C(C)C)=NC=C2Cl
Molecular Weight (g/mol) 414.89
Formula Weight 414.89
Percent Purity 98.11%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.