CAS RN 1203588-99-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Guanidinobenzoic Acid-D4 Hydrochloride, TRC
CAS: 1203588-99-1 Molecular Formula: C8H5D4N3O2 . HCl Molecular Weight (g/mol): 219.66 Synonym: 4-[(Aminoiminomethyl)amino]benzoic Acid Hydrochloride-D4,p-Guanidinobenzoic Acid Hydrochloride-D4 IUPAC Name: 4-guanidinobenzoic-2,3,5,6-d4 acid hydrochloride SMILES: [2H]C1=C(C(O)=O)C([2H])=C([2H])C(NC(N)=N)=C1[2H].Cl