missing translation for 'onlineSavingsMsg'
Learn More

4-Guanidinobenzoic Acid-D4 Hydrochloride, TRC

Product Code. 30406146
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30406146 25 mg 25mg
30389628 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30406146 Supplier Toronto Research Chemicals Supplier No. TRCG82130225MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1203588-99-1
Molecular Formula C8H5D4N3O2 . HCl
Molecular Weight (g/mol) 219.66
Synonym 4-[(Aminoiminomethyl)amino]benzoic Acid Hydrochloride-D4, p-Guanidinobenzoic Acid Hydrochloride-D4
IUPAC Name 4-guanidinobenzoic-2,3,5,6-d4 acid hydrochloride
SMILES [2H]C1=C(C(O)=O)C([2H])=C([2H])C(NC(N)=N)=C1[2H].Cl

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C8H9N3O2.ClH/c9-8(10)11-6-3-1-5(2-4-6)7(12)13;/h1-4H,(H,12,13)(H4,9,10,11);1H/i1D,2D,3D,4D;
Formula Weight 219.55
Chemical Name or Material 4-Guanidinobenzoic Acid-D4 Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.