CAS RN 1196070-26-4
CAS RN 1196070-26-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Fiboflapon sodium, MedChemExpress
MedChemExpress Fiboflapon sodium (GSK2190915; AM-803) is a potent and orally bioavailable 5-lipoxygenase-activating protein (FLAP) inhibitor with a potency of 2.9 nM in FLAP binding, an IC50 of 76 nM for inhibition of LTB4 in human blood.