missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fiboflapon sodium, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
Fiboflapon sodium (GSK2190915
Specifications
Specifications
| Chemical Name or Material | Fiboflapon sodium |
| CAS | 1196070-26-4 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C38H42N3NaO4S |
| Quantity | 50 mg |
| Synonym | GSK2190915 sodium salt AM-803 sodium |
| Solubility Information | DMSO : ≥ 32 mg/mL (48.50 mM) |
| SMILES | CC(C(O[Na])=O)(C)CC1=C(SC(C)(C)C)C2=CC(OCC3=CC=C(C)C=N3)=CC=C2N1CC4=CC=C(C5=CN=C(OCC)C=C5)C=C4 |
| Molecular Weight (g/mol) | 659.81 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction