CAS RN 118974-02-0
CAS RN 118974-02-0
IUPAC Name:
(1S,12S,15S)-7-methoxy-12-(2-methylprop-1-en-1-yl)-10,13,19-triazapentacyclo[11.7.0.0³,¹¹.0⁴,⁹.0¹⁵,¹⁹]icosa-3(11),4,6,8-tetraene-14,20-dione
Synonyms:
(1S,12S,15S)-7-methoxy-12-(2-methylprop-1-en-1-yl)-10,13,19-triazapentacyclo[11.7.0.0³,¹¹.0⁴,⁹.0¹⁵,¹⁹]icosa-3(11),4,6,8-tetraene-14,20-dione
Molecular Weight (g/mol):
379.46
Molecular Formula:
C22H25N3O3
InChi Key:
DBEYVIGIPJSTOR-FHWLQOOXSA-N
SMILES:
COC1=CC=C2C(NC3=C2C[C@@H]2N([C@H]3C=C(C)C)C(=O)[C@@H]3CCCN3C2=O)=C1
1
–
1
of
1
results
1
Fumitremorgin C, MedChemExpress
MedChemExpress Fumitremorgin C is a potent and selective ABCG2/BRCP inhibitor.