missing translation for 'onlineSavingsMsg'
Learn More

Fumitremorgin C, MedChemExpress

Product Code. 16412589
Encompass_Preferred
Click to view available options
Quantity:
1 mg
250 μg
Unit Size:
1mg
250µg
This item is not returnable. View return policy

Product Code. 16412589

Brand: Medchem Express HYN2143

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Fumitremorgin C is a potent and selective ABCG2/BRCP inhibitor.

Fumitremorgin C is a potent and selective ABCG2/BRCP inhibitor.

Specifications

Chemical Name or Material Fumitremorgin C
CAS 118974-02-0
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H25N3O3
Quantity 1 mg
Synonym 12α-Fumitremorgin C
Solubility Information DMSO : 50 mg/mL (131.77 mM; Need ultrasonic)
SMILES O=C([C@]1([H])CC2=C([C@H](/C=C(C)\C)N13)NC4=C2C=CC(OC)=C4)N5[C@](CCC5)([H])C3=O
Molecular Weight (g/mol) 379.45
Formula Weight 379.45
Percent Purity 98.26%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.