CAS RN 1189727-93-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Didesmethyl Sibutramine-d6, TRC
CAS: 1189727-93-2 Molecular Formula: C15H16D6ClN Molecular Weight (g/mol): 257.83 Synonym: 1-(4-Chlorophenyl)-alpha-(2-methylpropyl)cyclobutanemethanamine-d6,(+/-)-Didesmethylsibutramine-d6,1-[1-(4-Chlorophenyl)cyclobutyl-d6]-3-methylbutylamine,1-[1-(4-Chlorophenyl)cyclobutyl-d6]-3-methylbutan-1-amine SMILES: [2H]C1([2H])C([2H])([2H])C(C(N)CC(C)C)(c2ccc(Cl)cc2)C1([2H])[2H]