missing translation for 'onlineSavingsMsg'
Learn More

Didesmethyl Sibutramine-d6, TRC

Product Code. 30386989
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386989 10 mg
30370484 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30386989 Supplier Toronto Research Chemicals Supplier No. TRCD44116710MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189727-93-2
Molecular Formula C15H16D6ClN
Molecular Weight (g/mol) 257.83
Synonym 1-(4-Chlorophenyl)-alpha-(2-methylpropyl)cyclobutanemethanamine-d6, (+/-)-Didesmethylsibutramine-d6, 1-[1-(4-Chlorophenyl)cyclobutyl-d6]-3-methylbutylamine, 1-[1-(4-Chlorophenyl)cyclobutyl-d6]-3-methylbutan-1-amine
SMILES [2H]C1([2H])C([2H])([2H])C(C(N)CC(C)C)(c2ccc(Cl)cc2)C1([2H])[2H]

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H22ClN/c1-11(2)10-14(17)15(8-3-9-15)12-4-6-13(16)7-5-12/h4-7,11,14H,3,8-10,17H2,1-2H3/i3D2,8D2,9D2
Chemical Name or Material Didesmethyl Sibutramine-d6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.