accessibility menu, dialog, popup

UserName

CAS RN 1182284-46-3

CH3ONHBrOOCH3

CAS RN 1182284-46-3

IUPAC Name: methyl 2-(4-bromo-2-acetamidophenyl)acetate
Synonyms: methyl 2-(4-bromo-2-acetamidophenyl)acetate
Molecular Weight (g/mol): 286.13
Molecular Formula: C11H12BrNO3
InChi Key: LCQMFPAASSGQFD-UHFFFAOYSA-N
SMILES: COC(=O)CC1=C(NC(C)=O)C=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results