missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ P-3FAX-Neu5Ac
GREENER_CHOICE

Product Code. 18767088
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18767088 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18767088 Supplier Tocris Bioscience™ Supplier No. 5760/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sialic acid analog; sialyltransferase inhibitor

P-3FAX-Neu5Ac is a cell-permeable sialic acid analog. Deacetylation occurs intracellularly giving rise to a sialyltransferase inhibitor. Abolishes expression of SLex expression on HL-60 cells and reduces E-selectin and P-selectin binding. Prevents metastasis formation in a mouse lung metastasis model.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-(acetylamino)-3,5-dideoxy-3-fluoro-D-erythro-α-L-manno-2-nonulopyranosonic acid methyl ester 2,4,7,8,9-pentaacetate
CAS 117405-58-0
Quantity 10 mg
Target Other Transferase Inhibitors
Molecular Formula C22H30FNO14
Formula Weight 551.47
Purity >97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.