accessibility menu, dialog, popup

UserName

CAS RN 114878-60-3

ONONNHCl

CAS RN 114878-60-3

IUPAC Name: 1-(2-chloro-4-nitrophenyl)piperazine
Synonyms: 1-(2-chloro-4-nitrophenyl)piperazine
Molecular Weight (g/mol): 241.68
Molecular Formula: C10H12ClN3O2
InChi Key: JNUWBYPJOBSXDN-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(N2CCNCC2)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results