accessibility menu, dialog, popup

UserName

CAS RN 114872-98-9

ClClOHONH2

CAS RN 114872-98-9

IUPAC Name: 2-amino-3-(2,4-dichlorophenyl)propanoic acid
Synonyms: 2-amino-3-(2,4-dichlorophenyl)propanoic acid
Molecular Weight (g/mol): 234.08
Molecular Formula: C9H9Cl2NO2
InChi Key: GWHQTNKPTXDNRM-UHFFFAOYNA-N
SMILES: NC(CC1=C(Cl)C=C(Cl)C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)