CAS RN 1134-23-2
CAS RN 1134-23-2
IUPAC Name:
N-cyclohexyl-N-ethyl(ethylsulfanyl)formamide
Synonyms:
cycloate
Molecular Weight (g/mol):
215.36
Molecular Formula:
C11H21NOS
InChi Key:
DFCAFRGABIXSDS-UHFFFAOYSA-N
SMILES:
CCSC(=O)N(CC)C1CCCCC1
1
–
3
of
3
results
1
Cycloate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Cycloate, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality