missing translation for 'onlineSavingsMsg'
Learn More

Cycloate in Methanol 100μg/mL, Fisher Chemical™

Product Code. 12963244 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
12963244 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 12963244

Brand: Fisher Chemical P875020

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 1134-23-2
Molecular Formula C11H21NOS
Concentration 100 μg/mL
InChI Key DFCAFRGABIXSDS-UHFFFAOYSA-N
SMILES CCN(C1CCCCC1)C(=O)SCC
IUPAC Name S-ethyl N-cyclohexyl-N-ethylcarbamothioate
Solvent or Matrix Methanol
Molecular Weight (g/mol) 215.355
PubChem CID 14337
Quantity 1 mL
ChEBI CHEBI:81955
Chemical Name or Material Cycloate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1 ML Metabolite Cycloate in Methanol 100µg/mL 1mL
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
Supplemental information
  • MIXTURE LIST-Contains : methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.