missing translation for 'onlineSavingsMsg'
Learn More

Cycloate, TRC

Product Code. 30350427
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30350427 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350427 Supplier Toronto Research Chemicals Supplier No. TRCC9893205G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cycloate
CAS 1134-23-2
Molecular Formula C11 H21 N O S
Quantity 5 g
Synonym Carbamothioic acid, cyclohexylethyl-, S-ethyl ester (9CI), Cyclohexanecarbamic acid, N-ethylthio-, S-ethyl ester (7CI,8CI), Buranit, Cycloate, Ethyl N-cyclohexyl-N-ethylthiolcarbamate, Etsan, Eurex, Hexylthiocarbam, R 2063, Ro-Neet, Ro-Neet 6E, Roneet 72LE, Ronit, S-Ethyl N-cyclohexyl-N-ethylthiocarbamate, S-Ethyl N-cyclohexyl-N-ethylthiolcarbamate, S-Ethyl cyclohexylethylthiocarbamate, S-Ethyl-N-ethyl-N-cyclohexylthiolcarbamate, Sabet
InChI Formula InChI=1S/C11H21NOS/c1-3-12(11(13)14-4-2)10-8-6-5-7-9-10/h10H,3-9H2,1-2H3
SMILES CCSC(=O)N(CC)C1CCCCC1
IUPAC Name S-ethyl N-cyclohexyl-N-ethylcarbamothioate
Molecular Weight (g/mol) 215.36
Formula Weight 215.1344
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.