accessibility menu, dialog, popup

UserName

CH3NH3CH3COOOOHONNNNNH2OPOHOOPOHOOPOOHHO

CAS RN 112898-15-4

IUPAC Name: ({[({[5-(6-amino-9H-purin-9-yl)-3-(4-benzoylbenzoyloxy)-4-hydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)phosphonic acid; triethylamine
Synonyms: ({[5-(6-aminopurin-9-yl)-3-(4-benzoylbenzoyloxy)-4-hydroxyoxolan-2-yl]methoxy(hydroxy)phosphoryl}oxy(hydroxy)phosphoryl)oxyphosphonic acid; triethylamine
Molecular Weight (g/mol): 816.59
Molecular Formula: C30H39N6O15P3
InChi Key: HVOVBTNCGADRTH-UHFFFAOYNA-N
SMILES: CCN(CC)CC.NC1=C2N=CN(C3OC(COP(O)(=O)OP(O)(=O)OP(O)(O)=O)C(OC(=O)C4=CC=C(C=C4)C(=O)C4=CC=CC=C4)C3O)C2=NC=N1
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results