missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BzATP triethylammonium salt

Description
Prototypic P2X7 receptor agonist (EC50 values are 3.6, 7 and 285 μM for rat, human and mouse receptors respectively). Exhibits 5 - 10 fold greater potency than ATP. Exhibits partial agonist activity at P2X1 (pEC50 = 8.7) and P2Y1 receptors and can be used as a photoaffinity label for ATPase.
This compound is a mixture of isomers.
Specifications
Specifications
| CAS | 112898-15-4 |
| Synonym | 2' 3'-o-4-benzoylbenzoyl adenosine 5'-triphosphate triethylammonium salt |
| Quantity | 1mg, 1 mg |
| Molecular Formula | C30H39N6O15P3 |
| Formula Weight | 1018.97 |
| Chemical Name or Material | BzATP triethylammonium salt |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?