accessibility menu, dialog, popup

UserName

CAS RN 1072952-41-0

BNOHOOH

CAS RN 1072952-41-0

IUPAC Name: [2-(benzyloxy)pyridin-3-yl]boronic acid
Synonyms: 2-(benzyloxy)pyridin-3-ylboronic acid
Molecular Weight (g/mol): 229.04
Molecular Formula: C12H12BNO3
InChi Key: YTIGWZQYGHOXOB-UHFFFAOYSA-N
SMILES: OB(O)C1=C(OCC2=CC=CC=C2)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results