missing translation for 'onlineSavingsMsg'
Learn More

5-[[4-(Hydroxymethyl)phenoxy]methyl]-3-isopropyl-1,3-oxazolidin-2-one (Bisoprolol Oxazolidinonebenzylalcohol), Mikromol™

Product Code. 30050738
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050738 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050738 Supplier Mikromol Supplier No. MM0460.160025
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

5-[[4-(Hydroxymethyl)phenoxy]methyl]-3-isopropyl-1,3-oxazolidin-2-one (Bisoprolol Oxazolidinonebenzylalcohol)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-[(4-(Hydroxymethyl)phenoxy)methyl]-3-isopropyl-1,3-oxazolidin-2-one (Bisoprolol Oxazolidinonebenzylalcohol)
CAS 1071765-44-0
Analyte or Component Names 5-[(4-(Hydroxymethyl)phenoxy)methyl]-3-isopropyl-1,3-oxazolidin-2-one (Bisoprolol Oxazolidinonebenzylalcohol)
Shipping Condition Room Temperature
Molecular Formula C14 H19 N O4
Certifications/Compliance ISO 9001
Synonym 5-[[4-(Hydroxymethyl)phenoxy]methyl]-3-isopropyl-1,3-oxazolidin-2-one, Bisoprolol Fumarate Imp. U (EP), Bisoprolol Oxazolidinonebenzylalcohol, Bisoprolol Imp. U (EP), Bisoprolol Fumarate Impurity U, Bisoprolol Impurity U
InChI Formula InChI=1S/C14H19NO4/c1-10(2)15-7-13(19-14(15)17)9-18-12-5-3-11(8-16)4-6-12/h3-6,10,13,16H,7-9H2,1-2H3
SMILES CC(C)N1CC(COc2ccc(CO)cc2)OC1=O
IUPAC Name 5-[[4-(hydroxymethyl)phenoxy]methyl]-3-propan-2-yl-1,3-oxazolidin-2-one
Molecular Weight (g/mol) 265.3
Quantity 25 mg
Formula Weight 265.1314 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.