CAS RN 107091-89-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MedChemExpress Thiazole Orange
MedChemExpress Thiazole orange is an unsymmetrical cyanine dye which can be conjugated to oligonucleotides (ONs) to create fluorogenic hybridisation probes. Thiazole orange can be used for reticulocyte analysis.