CAS RN 1068-61-7
IUPAC Name:
λ²-lead(2+) bis(2-methylprop-2-enoate)
Synonyms:
λ²-lead(2+) dimethacrylate
Molecular Weight (g/mol):
377.4
Molecular Formula:
C8H10O4Pb
InChi Key:
FEAGCKGOSVNLMF-UHFFFAOYSA-L
SMILES:
[Pb++].CC(=C)C([O-])=O.CC(=C)C([O-])=O
1
–
1
of
1
results
1
Lead(II) methacrylate, 96%, Thermo Scientific™
CAS: 1068-61-7 Molecular Formula: C8H10O4Pb Molecular Weight (g/mol): 377.364 MDL Number: MFCD00080549 InChI Key: FEAGCKGOSVNLMF-UHFFFAOYSA-L Synonym: lead 2+ methacrylate,lead ii methacrylate,acmc-20aknj,2-propenoic acid, 2-methyl-, lead 2+ salt,2-propenoic acid, 2-methyl-, lead salt, basic,bismethacrylic acid lead ii salt,lead 2+ ; 2-methylprop-2-enoate,2-propenoic acid, 2-methyl-, lead 2+ salt 2:1 PubChem CID: 61255 IUPAC Name: lead(2+);2-methylprop-2-enoate SMILES: CC(=C)C(=O)[O-].CC(=C)C(=O)[O-].[Pb+2]