missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) methacrylate, 96%, Thermo Scientific™

Product Code. 11348688
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11348688 100 g 100g
11338688 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11348688 Supplier Thermo Scientific Alfa Aesar Supplier No. 041569.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1068-61-7
Molecular Formula C8H10O4Pb
Molecular Weight (g/mol) 377.364
MDL Number MFCD00080549
InChI Key FEAGCKGOSVNLMF-UHFFFAOYSA-L
Synonym lead 2+ methacrylate, lead ii methacrylate, acmc-20aknj, 2-propenoic acid, 2-methyl-, lead 2+ salt, 2-propenoic acid, 2-methyl-, lead salt, basic, bismethacrylic acid lead ii salt, lead 2+ ; 2-methylprop-2-enoate, 2-propenoic acid, 2-methyl-, lead 2+ salt 2:1
PubChem CID 61255
IUPAC Name lead(2+);2-methylprop-2-enoate
SMILES CC(=C)C(=O)[O-].CC(=C)C(=O)[O-].[Pb+2]

Specifications

Melting Point 62°C
Odor Odorless
Linear Formula [H2C=C(CH3)CO2]2Pb
Quantity 100 g
UN Number UN2291
Formula Weight 377.36
Percent Purity 96%
Physical Form Powder
Chemical Name or Material Lead(II) methacrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.