accessibility menu, dialog, popup

UserName

CAS RN 102582-93-4

FFFSOHO

CAS RN 102582-93-4

IUPAC Name: 2-{[4-(trifluoromethyl)phenyl]sulfanyl}acetic acid
Synonyms: {[4-(trifluoromethyl)phenyl]sulfanyl}acetic acid
Molecular Weight (g/mol): 236.21
Molecular Formula: C9H7F3O2S
InChi Key: MBVVWIKWEHKYGF-UHFFFAOYSA-N
SMILES: OC(=O)CSC1=CC=C(C=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results